Få mere at vide
Chlorquinaldol, LoGiCal
Artikelnummer. 30463000
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463000 10 mg
1 options
Artikelnummer. 30463000 Leverandør LoGiCal Leverandørnr. LGCFOR0390.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorquinaldol
CAS 72-80-0
Analyt- eller komponentnavne Chlorquinaldol
Forsendelsestilstand Room Temperature
Molekylær formel C10 H7 Cl2 N O
Certificeringer/compliance ISO 9001
Synonym Chlorquinaldol, 5,7-Dichloro-2-methyl-8-hydroxyquinoline, 5,7-Dichloro-2-methyl-8-quinolinol, 5,7-Dichloro-8-hydroxy-2-methylquinoline, 5,7-Dichloro-8-hydroxyquinaldine, 5,7-Dichloro-8-quinaldinol, Acnosan, Afungil, Chloquinan, Chlorchinaldin, Chlorchinaldol, Chlorguinaldon, Chlorquinaldol, Florabina, Gyno-Sterosan, Gynotherax, Hydroxydichloroquinaldinol, Intensol, Quesil, Saprosan, Siogen, Siogenal, Siogene, Siogeno, Siogenon, Siosept, Siosteran, Sterosan, Steroxin, Sterozan, Vagisteran
InChI formel InChI=1S/C10H7Cl2NO/c1-5-2-3-6-7(11)4-8(12)10(14)9(6)13-5/h2-4,14H,1H3
Løsningstype Neat
SMIL Cc1ccc2c(Cl)cc(Cl)c(O)c2n1
IUPAC navn 5,7-dichloro-2-methylquinolin-8-ol
Industry Type Clinical Testing
Molekylvægt (g/mol) 228.07
Mængde 10 mg
Formel vægt 226.9905 g/mol