Få mere at vide
Chlorpyriphos-methyl, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorpyrifos-methyl |
| CAS | 5598-13-0 |
| Molekylær formel | C7 H7 Cl3 N O3 P S |
| Mængde | 250 mg |
| Synonym | Phosphorothioic acid, O,O-dimethyl O-(3,5,6-trichloro-2-pyridyl) ester (7CI,8CI), Chlorpyrifos M, Chlorpyrifos-methyl, Chlorpyriphos-methyl, Cyfen, Dowco 214, Dursban ME, Dursban methyl, ENT 27520, M 3196, Methyl Dursban, Methyl chlorpyrifos, Noltran, O,O-Dimethyl O-(3,5,6-trichloro-2-pyridyl) phosphorothioate, O,O-Dimethyl O-(3,5,6-trichloro-2-pyridyl) thiophosphate, O-(3,5,6-Trichloro-2-pyridinyl) O,O-dimethyl phosphorothioate, OMS 1155, Reldan, Reldan E |
| InChI formel | InChI=1S/C7H7Cl3NO3PS/c1-12-15(16,13-2)14-7-5(9)3-4(8)6(10)11-7/h3H,1-2H3 |
| SMIL | COP(=S)(OC)Oc1nc(Cl)c(Cl)cc1Cl |
| IUPAC navn | dimethoxy-sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 322.53 |
| Formel vægt | 320.895 |