Få mere at vide
Chlorpyrifos Oxon, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorpyrifos-oxon |
| CAS | 5598-15-2 |
| Molekylær formel | C9 H11 Cl3 N O4 P |
| Mængde | 100 mg |
| Synonym | Phosphoric acid, diethyl 3,5,6-trichloro-2-pyridyl ester (7CI,8CI), 2-Pyridinol, 3,5,6-trichloro-, diethyl phosphate (ester) (8CI), 3,5,6-Trichloro-2-pyridyl diethyl phosphate, Chloropyrifos oxon, Chlorpyrifos oxon, Chlorpyrifos-ethyl oxon, Chlorpyrifoxon, Chlorpyriphoxon, Diethyl 3,5,6-trichloro-2-pyridyl phosphate, Dursban oxygen analog, Dursbanoxon, Fospirate-ethyl, O,O-Diethyl O-3,5,6-trichloro-2-pyridyl phosphate, O-(3,5,6-Trichloro-2-pyridinyl) O,O-diethyl phosphate |
| InChI formel | InChI=1S/C9H11Cl3NO4P/c1-3-15-18(14,16-4-2)17-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3 |
| SMIL | CCOP(=O)(OCC)Oc1nc(Cl)c(Cl)cc1Cl |
| IUPAC navn | diethyl (3,5,6-trichloropyridin-2-yl) phosphate |
| Molekylvægt (g/mol) | 334.52 |
| Formel vægt | 332.9491 |