missing translation for 'onlineSavingsMsg'
Få mere at vide

Chlorpyrifos Oxon-d10, TRC

Artikelnummer. 30358232
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358232 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358232 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4253221MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Chlorpyrifos Oxon-d10
CAS 1794779-85-3
Molekylær formel C9HD10Cl3NO4P
Mængde 1 mg
Synonym Phosphoric Acid (Diethyl-d10) 3,5,6-Trichloro-2-pyridinyl Ester, 6-Trichloro-2-pyridyl (Diethyl-d10) Phosphate, Chloropyrifos Oxon-d10, Chlorpyrifos(ethyl-d10) Oxon, Chlorpyriphoxon-d10, (Diethyl-d10) 3,5,6-Trichloro-2-pyridyl Phosphate, Dursban-d10 Oxygen Analog, Dursbanoxon-d10, Fospirate (ethyl-d10), O,O-(Diethyl-d10) O-3,5,6-Trichloro-2-pyridyl Phosphate, O-(3,5,6-Trichloro-2-pyridinyl) O,O-(Diethyl-d10) Phosphate
InChI formel InChI=1S/C9H11Cl3NO4P/c1-3-15-18(14,16-4-2)17-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2
SMIL ClC1=C(Cl)C=C(Cl)C(OP(OC([2H])([2H])C([2H])([2H])[2H])(OC([2H])([2H])C([2H])([2H])[2H])=O)=N1
IUPAC navn bis(1,1,2,2,2-pentadeuterioethyl) (3,5,6-trichloropyridin-2-yl) phosphate
Molekylvægt (g/mol) 344.58
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.