Få mere at vide
Chlorpyrifos Oxon-d10, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorpyrifos Oxon-d10 |
| CAS | 1794779-85-3 |
| Molekylær formel | C9HD10Cl3NO4P |
| Mængde | 10 mg |
| Synonym | Phosphoric Acid (Diethyl-d10) 3,5,6-Trichloro-2-pyridinyl Ester, 6-Trichloro-2-pyridyl (Diethyl-d10) Phosphate, Chloropyrifos Oxon-d10, Chlorpyrifos(ethyl-d10) Oxon, Chlorpyriphoxon-d10, (Diethyl-d10) 3,5,6-Trichloro-2-pyridyl Phosphate, Dursban-d10 Oxygen Analog, Dursbanoxon-d10, Fospirate (ethyl-d10), O,O-(Diethyl-d10) O-3,5,6-Trichloro-2-pyridyl Phosphate, O-(3,5,6-Trichloro-2-pyridinyl) O,O-(Diethyl-d10) Phosphate |
| InChI formel | InChI=1S/C9H11Cl3NO4P/c1-3-15-18(14,16-4-2)17-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMIL | ClC1=C(Cl)C=C(Cl)C(OP(OC([2H])([2H])C([2H])([2H])[2H])(OC([2H])([2H])C([2H])([2H])[2H])=O)=N1 |
| IUPAC navn | bis(1,1,2,2,2-pentadeuterioethyl) (3,5,6-trichloropyridin-2-yl) phosphate |
| Molekylvægt (g/mol) | 344.58 |