Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorpyrifos methyl |
| CAS | 5598-13-0 |
| CAS Max % | ≤ 100% |
| Beilstein | 1541078 |
| Farve | Colorless |
| Smeltepunkt | 45°C to 46°C |
| Synonym | O,O-dimethyl O-(3,5,6-trichloro-2-pyridinyl) phosphorothioate |
| Molekylvægt (g/mol) | 322.53 g/mol |
| Mængde | 100 mg |