Få mere at vide
Chlorprothixene EP Impurity B (as Hydrochloride), Mikromol™
Beskrivelse
Chlorprothixene EP Impurity B (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N,N-Dimethyl-3-(9H-thioxanthen-9-ylidene)propan-1-amine Hydrochloride |
| CAS | 4907-84-0 |
| Analyt- eller komponentnavne | N,N-Dimethyl-3-(9H-thioxanthen-9-ylidene)propan-1-amine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H19 N S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Chlorprothixene Hydrochloride Imp. B (EP), Chlorprothixene Imp. B (EP), N,N-Dimethyl-3-(9H-thioxanthen-9-ylidene)propan-1-amine Hydrochloride, Chlorprothixene Hydrochloride Imp. B (EP) as Hydrochloride, Chlorprothixene Hydrochloride Impurity B as Hydrochloride, Chlorprothixene Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C18H19NS.ClH/c1-19(2)13-7-10-14-15-8-3-5-11-17(15)20-18-12-6-4-9-16(14)18;/h3-6,8-12H,7,13H2,1-2H3;1H |
| SMIL | Cl.CN(C)CCC=C1c2ccccc2Sc3ccccc13 |
| IUPAC navn | N,N-dimethyl-3-thioxanthen-9-ylidenepropan-1-amine;hydrochloride |
| Vis mere |