Få mere at vide
Chlorprothixene-d6, TRC
Artikelnummer. 30350566
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30350566 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350566 Leverandør Toronto Research Chemicals Leverandørnr. TRCC42491210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorprothixene-d6
CAS 1246912-83-3
Molekylær formel C18H12D6ClNS
Mængde 10 mg
Synonym (3Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-1-propanamine-d6, (Z)-2-Chloro-N,N-dimethyl-thioxanthene-delta9,gamma-propylamine-d6, (Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-1-propanamine-d6, CPT-d6, 3-(2-Chlorothioxanthen-9-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine
InChI formel InChI=1S/C18H18ClNS/c1-20(2)11-5-7-14-15-6-3-4-8-17(15)21-18-10-9-13(19)12-16(14)18/h3-4,6-10,12H,5,11H2,1-2H3/b14-7-/i1D3,2D3
SMIL [2H]C([2H])([2H])N(CC\C=C/1\c2ccccc2Sc3ccc(Cl)cc13)C([2H])([2H])[2H]
IUPAC navn (3Z)-3-(2-chlorothioxanthen-9-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine
Molekylvægt (g/mol) 321.9