Få mere at vide
Chlorpropham-d7 (iso-propyl-d7), CDN
Artikelnummer. 30325647
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325647 0.01 g
30324052 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325647 Leverandør CDN Leverandørnr. CDND80630.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorpropham D7 (isopropyl D7)
CAS 2140327-49-5
Molekylær formel C10 D7 H5 Cl N O2
Mængde 0.01 g
Synonym Chlorpropham D7 (isopropyl D7)
InChI formel InChI=1 S/C10H12ClNO2/c1-7(2)14-10(13)12-9-5-3-4-8(11)6-9/h3-7 H,1-2H3,(H,12,13)/i1D3,2D3,7 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(OC(=O)Nc1cccc(Cl)c1)C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,3,3,3-heptadeuteriopropan-2-yl N-(3-chlorophenyl)carbamate
Molekylvægt (g/mol) 220.7039
Formel vægt 220.0996 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity