Få mere at vide
Chlorpropamide-d4, TRC
Artikelnummer. 30367851
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30367851 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367851 Leverandør Toronto Research Chemicals Leverandørnr. TRCC424802100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorpropamide-d4
CAS 1794779-79-5
Molekylær formel C10H9D4ClN2O3S
Mængde 100 mg
Synonym 4-Chloro-N-[(propylamino)carbonyl]benzenesulfonamide-d4, Adiaben-d4, Asucrol-d4, Catanil-d4, Chlorodiabina-d4, Chloronase-d4, Chloropropamide-d4, Chlorpropamid-d4, Diabaril-d4, Diabechlor-d4, Diabenal-d4, Diabenese-d4, Glisema-d4, Meldian-d4, Melitase-d4, Millinese-d4, N-(4-Chlorophenylsulfonyl)-N'-propylurea-d4, NSC 44634-d4, NSC 626720-d4, P 607-d4, Stabinol-d4, U 9818-d4, N-(p-Chlorobenzenesulfonyl)-N'-propylurea-d4, N-Propyl-N'-(p-chlorobenzenesulfonyl)urea-d4
InChI formel InChI=1S/C10H13ClN2O3S/c1-2-7-12-10(14)13-17(15,16)9-5-3-8(11)4-6-9/h3-6H,2,7H2,1H3,(H2,12,13,14)/i3D,4D,5D,6D
SMIL ClC1=C([2H])C([2H])=C(S(NC(NCCC)=O)(=O)=O)C([2H])=C1[2H]
IUPAC navn 1-(4-chloro-2,3,5,6-tetradeuteriophenyl)sulfonyl-3-propylurea
Molekylvægt (g/mol) 280.76