Få mere at vide
Chlorpromazine Sulfone N-Oxide Hydrochloride (Chlorpromazine N,S,S-Trioxide Hydrochloride), Mikromol
Beskrivelse
Chlorpromazine Sulfone N-Oxide Hydrochloride (Chlorpromazine N,S,S-Trioxide Hydrochloride), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorpromazine Sulfone N-Oxide Hydrochloride |
| CAS | 109724-46-1 |
| Analyt- eller komponentnavne | Chlorpromazine Sulfone N-Oxide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H19 Cl N2 O3 S . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, N,5,5-trioxide, hydrochloride (1:1), Phenothiazine, 2-chloro-10-(3-dimethylaminopropyl)-, N,5,5-trioxide, hydrochloride (6CI), Chlorpromazine Sulfone N-Oxide Hydrochloride |
| InChI formel | InChI=1S/C17H19ClN2O3S.ClH/c1-20(2,21)11-5-10-19-14-6-3-4-7-16(14)24(22,23)17-9-8-13(18)12-15(17)19;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.C[N+](C)([O-])CCCN1c2ccccc2S(=O)(=O)c3ccc(Cl)cc13 |
| Vis mere |