missing translation for 'onlineSavingsMsg'
Få mere at vide

Chlorpromazine Sulfone Hydrochloride, TRC

Product Code. 30355708
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30355708 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355708 Supplier Toronto Research Chemicals Supplier No. TRCC424760500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Chlorpromazine Sulfone Hydrochloride
CAS 18683-73-3
Molekylær formel C17 H19 Cl N2 O2 S . Cl H
Mængde 500 mg
Synonym 10H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, 5,5-dioxide, hydrochloride (1:1), Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]-, 5,5-dioxide, monohydrochloride (6CI,7CI,8CI), Chlorpromazine Sulfone Hydrochloride
InChI formel InChI=1S/C17H19ClN2O2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)23(21,22)17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H
SMIL Cl.CN(C)CCCN1c2ccccc2S(=O)(=O)c3ccc(Cl)cc13
IUPAC navn 3-(2-chloro-5,5-dioxophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
Molekylvægt (g/mol) 387.32
Formel vægt 386.0623
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.