Få mere at vide
Chlorpromazine-D3 0.1 mg/ml in Methanol, LoGiCal
Artikelnummer. 30461515
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30461515 Leverandør LoGiCal Leverandørnr. LGCAMP0420.8002
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorpromazine-D3
CAS 136765-28-1
Opløsningsmiddel eller Matrix Methanol
Molekylær formel C17 2H3 H16 Cl N2 S
Mængde 1 mL
Synonym 10H-Phenothiazine-10-propanamine, 2-chloro-N-methyl-N-(methyl-d3)- (9CI), Chlorpromazine-D3
InChI formel InChI=1S/C17H19ClN2S/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20/h3-4,6-9,12H,5,10-11H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])N(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13
IUPAC navn 3-(2-chlorophenothiazin-10-yl)-N-methyl-N-(trideuteriomethyl)propan-1-amine
Molekylvægt (g/mol) 321.88
Formel vægt 321.1146 g/mol