Få mere at vide
Chlorpromazine-13C,d3 Hydrochloride, TRC
Artikelnummer. 30350611
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350611 1 mg
30355709 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350611 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4247531MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1329612-87-4
Molekylær formel C1613CH17D3Cl2N2S
Molekylvægt (g/mol) 359.34
Synonym 2-Chloro-N,N-(dimethyl-13C,d3)-10H-phenothiazine-10-propanamine Hydrochloride, 2-Chloro-10-[3-(dimethylamino-13C,d3)propyl]phenothiazine Hydrochloride, Klorproman-13C,d3, Marazine-13C,d3, Norcozine-13C,d3, Torazina-13C,d3, Tranzene-13C,d3
IUPAC navn 3-(2-chlorophenothiazin-10-yl)-N-methyl-N-(trideuterio(113C)methyl)propan-1-amine;hydrochloride
SMIL ClC1=CC(N2CCCN([13C]([2H])([2H])[2H])C)=C(C=C1)SC3=C2C=CC=C3.Cl
Tekniske data
Kemisk navn eller materiale Chlorpromazine-13C,d3 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H/i1+1D3;