Få mere at vide
Chlorproethazine-d10 Hydrochloride, TRC
Artikelnummer. 30368662
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30368662 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368662 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4247022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorproethazine-d10 Hydrochloride
CAS 1216730-87-8
Molekylær formel C19H14D10Cl2N2S
Mængde 2.5 mg
Synonym 2-Chloro-N,N-(diethyl-d10)-10H-phenothiazine-10-propanamine Hydrochloride, 2-Chloro-10-[3-(diethylamino-d10)propyl]phenothiazine Hydrochloride, Neuriplege-d10, RP 4909-d10
InChI formel InChI=1S/C19H23ClN2S.ClH/c1-3-21(4-2)12-7-13-22-16-8-5-6-9-18(16)23-19-11-10-15(20)14-17(19)22;/h5-6,8-11,14H,3-4,7,12-13H2,1-2H3;1H/i1D3,2D3,3D2,4D2;
SMIL Cl.[2H]C([2H])([2H])C([2H])([2H])N(CCCN1c2ccccc2Sc3ccc(Cl)cc13)C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 3-(2-chlorophenothiazin-10-yl)-N,N-bis(1,1,2,2,2-pentadeuterioethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 393.44
Formel vægt 392.1665