Få mere at vide
Chlorphenoxamine Hydrochloride, Mikromol™
Beskrivelse
Chlorphenoxamine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorphenoxamine Hydrochloride |
| CAS | 562-09-4 |
| Analyt- eller komponentnavne | Chlorphenoxamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H22 Cl N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Chlorphenoxamine Hydrochloride, 2-[1-(4-Chlorophenyl)-1-phenylethoxy]-N,N-dimethyl-ethanamine hydrochloride, Chlorphenoxamine hydrochloride, Clorevan, Contristamine, Contristamine hydrochloride, Phenoxene, Phenoxene hydrochloride, Phenoxine, Systral, 2-(p-Chloro-alpha-methyl-alpha-phenylbenzyloxy)-N,N-dimethylethylamine hydrochloride, [1-(p-Chlorophenyl)-1-phenyl]ethyl (beta-dimethylaminoethyl) ether hydrochloride, beta-Dimethylaminoethyl (p-chloro-alpha-methylbenzhydryl) ether hydrochloride |
| InChI formel | InChI=1S/C18H22ClNO.ClH/c1-18(21-14-13-20(2)3,15-7-5-4-6-8-15)16-9-11-17(19)12-10-16;/h4-12H,13-14H2,1-3H3;1H |
| SMIL | Cl.CN(C)CCOC(C)(c1ccccc1)c2ccc(Cl)cc2 |
| IUPAC navn | 2-[1-(4-chlorophenyl)-1-phenylethoxy]-N,N-dimethylethanamine;hydrochloride |
| Vis mere |