Få mere at vide
Chlorphenoxamide, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorphenoxamide |
| CAS | 3576-64-5 |
| Analyt- eller komponentnavne | Chlorphenoxamide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H16 Cl2 N2 O5 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Acetamide, 2,2-dichloro-N-(2-hydroxyethyl)-N-[p-(p-nitrophenoxy)benzyl]- (6CI,7CI,8CI), 2,2-Dichloro-N-2-hydroxyethyl-N-[p-(p-nitrophenoxy)benzyl]acetamide, Chlorophenoxamide, Chlorphenoxamide, Clefamide, Mebinol, Moebinol, Acetamide, 2,2-dichloro-N-(2-hydroxyethyl)-N-[[4-(4-nitrophenoxy)phenyl]methyl]- |
| InChI formel | InChI=1S/C17H16Cl2N2O5/c18-16(19)17(23)20(9-10-22)11-12-1-5-14(6-2-12)26-15-7-3-13(4-8-15)21(24)25/h1-8,16,22H,9-11H2 |
| Løsningstype | Neat |
| SMIL | OCCN(Cc1ccc(Oc2ccc(cc2)[N+](=O)[O-])cc1)C(=O)C(Cl)Cl |
| Vis mere |