Få mere at vide
Chloroxylenol-d6, TRC
Artikelnummer. 30365103
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30365103 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365103 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4279522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chloroxylenol-d6
CAS 1407521-66-7
Renhedsgrad noter HPLC
Molekylær formel C8H3D6ClO
Mængde 2.5 mg
Synonym 4-Chloro-3,5-dimethylphenol-d6, 4-Chloro-3,5-xylenol-d6, 2,6-Dimethyl-4-hydroxy-1-chlorobenzene-d6, 2-Chloro-5-hydroxy-1,3-dimethylbenzene-d6, 2-Chloro-5-hydroxy-m-xylene-d6, 4-Chloro-5,3-dimethylphenol-d6, 4-Chloro-m-xylenol-d6, Benzytol-d6, Camel-d6, Chloroxylenol-d6, Desson-d6, Dettol-d6, Ecotru-d6, Espadol-d6, Finecide C 2000-d6, Hokubarine E 400-d6, Husept Extra-d6, NSC 4971-d6, Nipacide PX-d6, Ottasept-d6, Ottasept Extra-d6, PCMX-d6, Parametaxylenol-d6, RBA 777-d6, p-Chloro-3,5-xylenol-d6, p-Chloro-m-Xylenol-d6
InChI formel InChI=1S/C8H9ClO/c1-5-3-7(10)4-6(2)8(5)9/h3-4,10H,1-2H3/i1D3,2D3
SMIL OC1=CC(C([2H])([2H])[2H])=C(Cl)C(C([2H])([2H])[2H])=C1
IUPAC navn 4-chloro-3,5-bis(trideuteriomethyl)phenol
Molekylvægt (g/mol) 162.65
Procent renhed >95