Learn More
chlorotripropylsilane, 98%, Thermo Scientific Chemicals
Catalog Number p-4258909
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
995-25-5
C9H21ClSi
192.81
ACTAPAGNZPZLEF-UHFFFAOYSA-N
tri-n-propylchlorosilane, tripropylchlorosilane, silane, chlorotripropyl, chlorotri-n-propylsilane, chloro tripropyl silane, chloro-tri-n-propylsilane, chlorotripropylsilane, chloro tri-n-propyl silane
70445
chloro(tripropyl)silane
CCC[Si](CCC)(CCC)Cl
995-25-5
C9H21ClSi
192.81
ACTAPAGNZPZLEF-UHFFFAOYSA-N
tri-n-propylchlorosilane, tripropylchlorosilane, silane, chlorotripropyl, chlorotri-n-propylsilane, chloro tripropyl silane, chloro-tri-n-propylsilane, chlorotripropylsilane, chloro tri-n-propyl silane
70445
chloro(tripropyl)silane
CCC[Si](CCC)(CCC)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8820g/mL
Boiling Point 199.0°C to 200.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3990 to 1.4410 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.882
Formula Weight 192.81
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material chlorotripropylsilane
Show More Show Less
compliance-icons
Product Identifier
  • Chlorotripropylsilane
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.