Få mere at vide
Chlorotrimethyl-d9-silane, CDN
Artikelnummer. 30312090
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312090 0.25 g -
30321777 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312090 Leverandør CDN Leverandørnr. CDND09530.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorotrimethylsilane-d9
CAS 20395-57-7
Molekylær formel C3 D9 Cl Si
Mængde 0.25 g
Synonym Chlorotrimethyl-d9-silane, Chlorotri(methyl-d3)silane, chloro-tris(trideuteriomethyl)silane
InChI formel InChI=1 S/C3H9ClSi/c1-5(2,3)4/h1-3H3/i1D3,2D3,3D3
SMIL [2 H]C([2 H])([2 H])[Si](Cl)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn chloro-tris(trideuteriomethyl)silane
Molekylvægt (g/mol) 117.7
Formel vægt 117.0727 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity