Få mere at vide
Chlorotrianisene-d9 (trimethoxy-d9), CDN
Artikelnummer. 30322119
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322119 0.01 g
30317395 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322119 Leverandør CDN Leverandørnr. CDND71980.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorotrianisene-D9 (trimethoxy-D9)
CAS 1276197-26-2
Molekylær formel C23 D9 H12 Cl O3
Mængde 0.01 g
Synonym Chlorotrianisene-d9, Chlorotrianisene-d9 (trimethoxy-d9), Chlorotrianisene-D9 (trimethoxy-D9), Chlorotris(p-methoxy-d3-phenyl)ethylene, Tri-p-anisylchloroethylene-d9, 1,1',1''-(1-chloro-1-ethenyl-2-ylidene)tris[4-(methoxy-d3)benzene]
InChI formel InChI=1 S/C23H21ClO3/c1-25-19-10-4-16(5-11-19)22(17-6-12-20(26-2)13-7-17)23(24)18-8-14-21(27-3)15-9-18/h4-15 H,1-3H3/i1D3,2D3,3D3
SMIL [2 H]C([2 H])([2 H])Oc1ccc(cc1)C(=C(c2ccc(OC([2 H])([2 H])[2 H])cc2)c3ccc(OC([2 H])([2 H])[2 H])cc3)Cl
IUPAC navn 1-[1-chloro-2,2-bis[4-(trideuteriomethoxy)phenyl]ethenyl]-4-(trideuteriomethoxy)benzene
Molekylvægt (g/mol) 389.9195
Formel vægt 389.1744 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity