Få mere at vide
Chlorothalonil, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorothalonil |
| CAS | 1897-45-6 |
| Analyt- eller komponentnavne | Chlorothalonil |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C8 Cl4 N2 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Isophthalonitrile, tetrachloro- (7CI,8CI), 2,4,5,6-Tetrachloro-1,3-benzenedicarbonitrile, 1,3-Dicyanotetrachlorobenzene, 2,4,5,6-Tetrachloro-1,3-dicyanobenzene, 2,4,5,6-Tetrachloro-1,3-isophthalonitrile, 2,4,5,6-Tetrachloro-m-benzenedinitrile, 2,4,5,6-Tetrachloroisophthalodinitrile, 2,4,5,6-Tetrachloroisophthalonitrile, Agronil PI 231, Barrack, Blotch, Bravo, Bravo 500, Bravo 500SC, Bravo 6F, Bravo 720, Bravo 72SC, Bravo Plus, Bravo Ultrex, Bravo Weather Stick, Bravo Weather Stik, Bravonil, Bravonil Ultrex, Busan 1192, Chloronil, Chloronil 720, Chlorthalonil, Clortosip, Clortosip 500SC, Clortosip L, DAC 2787, Daconil, Daconil 1000, Daconil 2787, Daconil 2787WP, Daconil 500, Daconil 50SC, Daconil F, Daconil M, Daconil Ultrex, Daconil Weather Stik, Daconil Weatherstik, Dacosoil, Dakoflo, Fongil F, Forturf, Fungitrol 404D, Guarant 500SC, Gwarant, Gwarant 500SC, Hortyl, Kabiguard 164SA, Kavach, Kavach 75WP, M 1, M 1 (mildewcide), Marincide C, Microban Additive M 15, Microban M 15, N 96, NexGen, Nopcocide, Nopcocide 54DB, Nopcocide N 54D, Nopcocide N 96, Nuocide 404D, Nuocide 960, Optimist, SD 404, Sweep, TPN, TPN (pesticide), Termil, Tetrachlorobenzene-1,3-dicarbonitrile, Tetrachloroisophthalonitrile, Thalonil, Tuff-Brite, Vanox, Vanox 500SC, Vanox 750PM, YS 801, YS 801 (mildewcide) |
| InChI formel | InChI=1S/C8Cl4N2/c9-5-3(1-13)6(10)8(12)7(11)4(5)2-14 |
| SMIL | Clc1c(Cl)c(C#N)c(Cl)c(C#N)c1Cl |
| IUPAC navn | 2,4,5,6-tetrachlorobenzene-1,3-dicarbonitrile |
| Vis mere |