Learn More
Chloropentaammineruthenium(III) chloride, Ru 33.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Chloropentaammineruthenium(III) chloride active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Orange-Yellow |
Linear Formula | [RuCl(NH3)5]Cl2 |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 292.58 |
Concentration or Composition (by Analyte or Components) | Ru 33.5% min. |
Physical Form | Micro Crystals |
Chemical Name or Material | Chloropentaammineruthenium(III) chloride |