Få mere at vide
Chloromethyl Isopropyl Carbonate, TRC
Artikelnummer. 30135264
Skift visning
Mængde:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135264 100 g
30010956 10 g
30007706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135264 Leverandør Toronto Research Chemicals Leverandørnr. TRCC369230100G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35180-01-9
Molekylær formel C5 H9 Cl O3
Molekylvægt (g/mol) 152.58
Synonym Carbonic acid, chloromethyl 1-methylethyl ester (9CI, ACI), Chloromethyl 1-methylethyl carbonate (ACI), [(Isopropyloxycarbonyl)oxy]methyl chloride, Chloromethyl isopropyl carbonate, Chloromethyl propan-2-yl carbonate, Isopropoxycarbonyloxymethyl chloride, Isopropyl chloromethyl carbonate
IUPAC navn chloromethyl propan-2-yl carbonate
SMIL CC(C)OC(=O)OCCl
Tekniske data
Kemisk navn eller materiale Chloromethyl Isopropyl Carbonate
Mængde 100 g
InChI formel InChI=1S/C5H9ClO3/c1-4(2)9-5(7)8-3-6/h4H,3H2,1-2H3
Formel vægt 152.024