Få mere at vide
Chloromethyl Isobutyrate, TRC
Artikelnummer. 30356782
Skift visning
Mængde:
250 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356782 250 mg
30359803 1 g
30352651 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30356782 Leverandør Toronto Research Chemicals Leverandørnr. TRCC381950250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61644-18-6
Molekylær formel C5H9O2Cl
Molekylvægt (g/mol) 136.57
Synonym (Isobutyryloxy)methyl chloride, Isobutyric acid chloromethyl ester, Chloromethyl 2-methylpropionate, Chloromethyl 2-methylpropanoate
IUPAC navn chloromethyl 2-methylpropanoate
SMIL CC(C)C(=O)OCCl
Tekniske data
Kemisk navn eller materiale Chloromethyl Isobutyrate
Mængde 250 mg
InChI formel InChI=1S/C5H9ClO2/c1-4(2)5(7)8-3-6/h4H,3H2,1-2H3
Formel vægt 136.0291