Få mere at vide
Chloroiodomethane-d2, CDN
Artikelnummer. 30309486
Skift visning
Mængde:
0.5 g
Pakningsstørrelse:
0.5g
Artikelnummer. Mængde unitSize
30309486 0.5 g 0.5g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309486 Leverandør CDN Leverandørnr. CDND52510.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chloroiodomethane-d2
CAS 129933-14-8
Molekylær formel C D2 Cl I
Mængde 0.5 g
Synonym Iodochloromethane-d2
InChI formel InChI=1 S/CH2ClI/c2-1-3/h1H2/i1D2
SMIL [2 H]C([2 H])(Cl)I
IUPAC navn chloro-dideuterio-iodo-methane
Molekylvægt (g/mol) 178.4
Formel vægt 177.9015 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity