Få mere at vide
Chlorocholine Chloride-d9, TRC
Artikelnummer. 30362009
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30362009 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362009 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36494850MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlormequat-chloride methyl-d9
CAS 1219257-11-0
Molekylær formel C5 2H9 H4 Cl N . Cl
Mængde 50 mg
Synonym Chlorocholine Chloride-d9, 2-Chloroethyl-tris(trideuteriomethyl)azanium chloride, Chlormequat-chloride (methyl-D9)
InChI formel InChI=1S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1/i1D3,2D3,3D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](CCCl)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2-chloroethyl-tris(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 167.12
Formel vægt 166.099