Tekniske data
| Kemisk navn eller materiale | γ-Chlorobutyric Acid |
| CAS | 627-00-9 |
| Molekylær formel | C4H7ClO2 |
| Mængde | 10 g |
| Synonym | 4-chlorobutyric acid, butanoic acid, 4-chloro, butyric acid, 4-chloro, gamma-chlorobutyric acid, 4-chlorobutanoicacid, gammachlorobutyric acid, 4-chloro-butyric acid, 4-chloro-n-butyric acid, .gamma.-chlorobutyric acid, 4-chloranylbutanoic acid |
| InChI nøgle | IPLKGJHGWCVSOG-UHFFFAOYSA-N |
| SMIL | C(CC(=O)O)CCl |
| IUPAC navn | 4-chlorobutanoic acid |
| Molekylvægt (g/mol) | 122.548 |
| PubChem CID | 12300 |
| Vis mere |