Få mere at vide
Chloroborane Methyl Sulfide Complex (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chloroborane Methyl Sulfide Complex (Technical Grade) |
| CAS | 63348-81-2 |
| Molekylær formel | C2H8BClS |
| Mængde | 1 g |
| Synonym | (T-4)-Chlorodihydro[1,1'-thiobis[methane]]boron, Chloroborane compd. with thiobis[methane], (T-4)-Chlorodihydro[thiobis[methane]]boron, Thiobismethane Boron Complex, Thiobismethane compd. with Chloroborane, Chloro(dimethyl sulfide)borane, Chloro(dimethyl sulfide)dihydroboron, Chloroborane-methyl Sulfide 1:1 Complex, Monochloroborane-dimethyl Sulfide |
| InChI formel | InChI=1S/C2H8BClS/c1-5(2)3-4/h3H2,1-2H3 |
| SMIL | C[S+](C)[BH2-]Cl |
| IUPAC navn | chloro(dimethylsulfonio)boranuide |
| Molekylvægt (g/mol) | 110.41 |
| Formel vægt | 110.0128 |