missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).
Solubility
Reacts with water.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).
Solubility
Reacts with water.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
Specifications
Boiling Point | 110°C (0.4 mmHg) |
Quantity | 1 g |
UN Number | UN3265 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 356.64 |
Percent Purity | 97% |
Chemical Name or Material | Chlorobis[4-(trifluoromethyl)phenyl]phosphine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'