missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%, Alfa Aesar™

Brand:  Alfa Aesar H25888.03

Additional Details : CAS Number : 13685-24-0 Weight : 0.00100kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11495630

  • 609.00 DKK / 1g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
Specifications

Chemical Identifiers

13685-24-0
356.632
PBLUOOURLNAOAJ-UHFFFAOYSA-N
4379933
C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)Cl
C14H8ClF6P
MFCD04972304
bis 4-trifluoromethylphenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphine, bis 4-trifluoromethyl phenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphane, acmc-20alnb, chloro-bis 4-trifluoromethyl phenyl phosphane
chloro-bis[4-(trifluoromethyl)phenyl]phosphane

Specifications

Chlorobis[4-(trifluoromethyl)phenyl]phosphine
110°C (0.4mmHg)
13685-24-0
MFCD04972304
bis 4-trifluoromethylphenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphine, bis 4-trifluoromethyl phenyl chlorophosphine, chlorobis 4-trifluoromethyl phenyl phosphane, acmc-20alnb, chloro-bis 4-trifluoromethyl phenyl phosphane
PBLUOOURLNAOAJ-UHFFFAOYSA-N
chloro-bis[4-(trifluoromethyl)phenyl]phosphane
4379933
97%
Air and moisture sensitive
1g
C14H8ClF6P
UN3265
Reacts with water.
C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)Cl
356.632
356.64
Videos
SDS Documents