Få mere at vide
Chlorobenzene-4-d1, CDN
Artikelnummer. 30322011
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322011 0.5 g 0.5g
30309434 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322011 Leverandør CDN Leverandørnr. CDND06970.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorobenzene-4-D
CAS 13122-34-4
Molekylær formel C6 D H4 Cl
Mængde 0.5 g
Synonym Benzene-d, 4-chloro- (9 CI, ACI), 4-Chlorobenzene-d (ACI), Benzene-d, p-chloro- (7 CI, 8 CI), Chlorobenzene-4-d, Chlorobenzene-4-D
InChI formel InChI=1 S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5 H/i1D
SMIL [2 H]c1ccc(Cl)cc1
IUPAC navn 1-chloro-4-deuteriobenzene
Molekylvægt (g/mol) 113.5631
Formel vægt 113.0143 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity