Få mere at vide
Chloroacetaldehyde Sodium Bisulfite, TRC
Artikelnummer. 30357636
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30357636 250 mg -
30356352 500 mg -
30358302 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30357636 Leverandør Toronto Research Chemicals Leverandørnr. TRCC382483250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13064-50-1
Molekylær formel C2H4ClNaO4S
Molekylvægt (g/mol) 182.56
Synonym Ethanesulfonic acid, 2-chloro-1-hydroxy-, sodium salt (1:1) (ACI), Ethanesulfonic acid, 2-chloro-1-hydroxy-, monosodium salt (8CI, 9CI), Ethanesulfonic acid, 2-chloro-1-hydroxy-, sodium salt (7CI), Monochloroacetaldehyde sodium bisulfite adduct, Sodium 2-chloro-1-hydroxyethane-1-sulfonate, Sodium 2-chloro-1-hydroxyethanesulfonate, Sodium monochloroacetaldehyde bisulfite, Chloroacetaldehyde sodium bisulfite
IUPAC navn sodium;2-chloro-1-hydroxyethanesulfonate
SMIL [Na+].OC(CCl)S(=O)(=O)[O-]
Tekniske data
Kemisk navn eller materiale Chloroacetaldehyde Sodium Bisulfite
Mængde 250 mg
InChI formel InChI=1S/C2H5ClO4S.Na/c3-1-2(4)8(5,6)7;/h2,4H,1H2,(H,5,6,7);/q;+1/p-1
Formel vægt 181.9417