Learn More
Chloro(triphenylphosphine)gold(I), 98%, BLD Pharm™
Specifications
Specifications
| CAS | 14243-64-2 |
| Molecular Formula | C18H15AuClP |
| MDL Number | MFCD00009588 |
| Quantity | 5 g |
| SMILES | [Au]Cl.P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
| Molecular Weight (g/mol) | 494.71 |
| Percent Purity | 98% |
| Chemical Name or Material | Chloro(triphenylphosphine)gold(I) |