missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Cyclopentadienylbis(triphenylphosphine)ruthenium Chloride is used as an efficient transfer hydrogenation catalyst. It is used in the formation of indoles, isoquinolines and quinolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 135°C (decomposition) |
Color | Orange |
Quantity | 1 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in water. |
IUPAC Name | chlororuthenium;cyclopentane;triphenylphosphane |
Formula Weight | 772.27 |
Physical Form | Crystalline Solid |
Chemical Name or Material | Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'