Få mere at vide
Chloro 2,3,5-Tri-O-p-chlorobenzoyl-Beta-D-ribofuranoside, TRC
Artikelnummer. 30364928
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364928 100 mg
30355950 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364928 Leverandør Toronto Research Chemicals Leverandørnr. TRCC421730100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2378379-44-1
Molekylær formel C26H18Cl4O7
Molekylvægt (g/mol) 584.23
Synonym β-D-Ribofuranosyl Chloride Tris(4-chlorobenzoate)
IUPAC navn (2S,3R,4R,5R)-2-chloro-5-(((4-chlorobenzoyl)oxy)methyl)tetrahydrofuran-3,4-diyl bis(4-chlorobenzoate)
SMIL Cl[C@@H]1O[C@H](COC(C2=CC=C(Cl)C=C2)=O)[C@@H](OC(C3=CC=C(Cl)C=C3)=O)[C@H]1OC(C4=CC=C(Cl)C=C4)=O
Tekniske data
Kemisk navn eller materiale Chloro 2,3,5-Tri-O-p-chlorobenzoyl-beta-D-ribofuranoside
Mængde 100 mg
InChI formel InChI=1S/C26H18Cl4O7/c27-17-7-1-14(2-8-17)24(31)34-13-20-21(36-25(32)15-3-9-18(28)10-4-15)22(23(30)35-20)37-26(33)16-5-11-19(29)12-6-16/h1-12,20-23H,13H2/t20-,21-,22-,23-/m1/s1
Formel vægt 581.98