Få mere at vide
Chlormequat-d9 Chloride (N,N,N-trimethyl-d9), CDN
Artikelnummer. 30317808
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317808 0.01 g
30311737 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317808 Leverandør CDN Leverandørnr. CDND77080.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlormequat-chloride methyl-d9
CAS 1219257-11-0
Molekylær formel C5 2H9 H4 Cl N . Cl
Mængde 0.01 g
Synonym Chlorocholine Chloride-d9, 2-Chloroethyl-tris(trideuteriomethyl)azanium chloride, Chlormequat-chloride (methyl-D9)
InChI formel InChI=1 S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1 H/q+1;/p-1/i1D3,2D3,3D3;
SMIL [Cl-].[2 H]C([2 H])([2 H])[N+](CCCl)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2-chloroethyl-tris(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 167.12
Formel vægt 166.099 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity