Få mere at vide
Chlorine Blank - Cl @ 0 μg/g in 75 cSt Mineral Oil, VHG™
Artikelnummer. 17950746
Skift visning
Mængde:
100 ml
500 ml
1 l
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17950746 500 ml
17911786 1 l
17911556 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17950746 Leverandør VHG Labs Leverandørnr. VHGCLOILBLK500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Chlorine Blank - Cl @ 0 μg/g in 75 cSt Mineral Oil
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorine
CAS 7782-50-5
Analyt- eller komponentnavne Chlorine
Forsendelsestilstand Room Temperature
Molekylær formel Cl2
Koncentration 0 μg/g
Certificeringer/compliance ISO 17034
Synonym Cl
InChI formel InChI=1S/Cl2/c1-2
SMIL ClCl
IUPAC navn molecular chlorine
Opløsningsmiddel eller Matrix Heavy Mineral Oil
Molekylvægt (g/mol) 70.91
Mængde 500 ml
Formel vægt 69.9377 g/mol