Få mere at vide
Chloridazon-d5 (Major), TRC
Artikelnummer. 30350302
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30350302 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350302 Leverandør Toronto Research Chemicals Leverandørnr. TRCC34000310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chloridazon phenyl-d5
CAS 1246818-99-4
Molekylær formel C10 2H5 H3 Cl N3 O
Mængde 10 mg
Synonym Chloridazon D5 (phenyl D5)
InChI formel InChI=1S/C10H8ClN3O/c11-9-8(12)6-13-14(10(9)15)7-4-2-1-3-5-7/h1-6H,12H2/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(N2N=CC(=C(Cl)C2=O)N)c([2H])c1[2H]
IUPAC navn 5-amino-4-chloro-2-(2,3,4,5,6-pentadeuteriophenyl)pyridazin-3-one
Molekylvægt (g/mol) 226.67
Formel vægt 226.067