Få mere at vide
Chlorhexidine Diacetate Impurity C, TRC
Artikelnummer. 30356211
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356211 50 mg
30362906 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356211 Leverandør Toronto Research Chemicals Leverandørnr. TRCC33500050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1'-[Hexane-1,6-diylbis(imino-carbonimidoyl)]bis[3-(4-chlorophenyl)urea]
CAS 62247-48-7
Molekylær formel C22 H28 Cl2 N8 O2
Mængde 50 mg
Synonym 2,4,11,13-Tetraazatetradecanediamide, N1,N14-bis(4-chlorophenyl)-3,12-diimino-, 2,4,11,13-Tetraazatetradecanediamide, N,N'-bis(4-chlorophenyl)-3,12-diimino- (9CI), N1,N14-Bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediamide
InChI formel InChI=1S/C22H28Cl2N8O2/c23-15-5-9-17(10-6-15)29-21(33)31-19(25)27-13-3-1-2-4-14-28-20(26)32-22(34)30-18-11-7-16(24)8-12-18/h5-12H,1-4,13-14H2,(H4,25,27,29,31,33)(H4,26,28,30,32,34)
SMIL Clc1ccc(NC(=O)NC(=N)NCCCCCCNC(=N)NC(=O)Nc2ccc(Cl)cc2)cc1
IUPAC navn 1-(4-chlorophenyl)-3-[N-[6-[[N-[(4-chlorophenyl)carbamoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]urea
Molekylvægt (g/mol) 507.4161
Formel vægt 506.1712