Få mere at vide
Chlorfenvinphos, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorfenvinphos |
| CAS | 470-90-6 |
| Molekylær formel | C12 H14 Cl3 O4 P |
| Mængde | 50 mg |
| Synonym | Benzyl alcohol, 2,4-dichloro-α-(chloromethylene)-, diethyl phosphate (6CI), Phosphoric acid, 2-chloro-1-(2,4-dichlorophenyl)vinyl diethyl ester (8CI), 2,4-Dichloro-α-(chloromethylene)benzyldiethyl phosphate, 2-Chloro-1-(2,4-dichlorophenyl)vinyl diethyl phosphate, Birlan, Birlane, Birlane 10G, Birlane Z 110, CVP, CVP (pesticide), Chlorfenvinfos, Chlorfenvinphos, Chlorphenvinfos, Chlorphenvinphos, Clofenvinfos, Compound 4072, Defiance S, Dermaton, Diethyl 1-(2,4-dichlorophenyl)-2-chlorovinyl phosphate, Diethyl 2-chloro-1-(2,4-dichlorophenyl)vinyl phosphate, ENT 24969, Enolofos, GC 4072, Haptasol, O,O-Diethyl O-[1-(2,4-dichlorophenyl)-2-chlorovinyl] phosphate, O,O-Diethyl O-[2-chloro-1-(2,4-dichlorophenyl)vinyl] phosphate, OMS 1328, Quirlan, SD 7859, Sapecron, Sapecron 50EC, Supona, Tarene, Unitox, Vinyphate, Z 110, Zaprawa enolofos |
| InChI formel | InChI=1S/C12H14Cl3O4P/c1-3-17-20(16,18-4-2)19-12(8-13)10-6-5-9(14)7-11(10)15/h5-8H,3-4H2,1-2H3 |
| SMIL | CCOP(=O)(OCC)OC(=CCl)c1ccc(Cl)cc1Cl |
| IUPAC navn | [2-chloro-1-(2,4-dichlorophenyl)ethenyl] diethyl phosphate |
| Molekylvægt (g/mol) | 359.57 |
| Formel vægt | 357.9695 |