Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Chlorfenvinfos | 470-90-6 | 1000μg/mL |
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlorfenvinfos |
| Emballage | 2mL Amber Prescored Glass Ampule |
| CAS | 470-90-6, 67-64-1 |
| CAS min % | 0.1, 99.9 |
| Koncentration eller sammensætningsnoter | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| CAS Max % | 0.1, 99.9 |
| Molekylær formel | C12H14Cl3O4P |
| Til brug med (udstyr) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| InChI nøgle | FSAVDKDHPDSCTO-XYOKQWHBSA-N |
| SMIL | CCOP(=O)(OCC)OC(=CCl)C1=C(C=C(C=C1)Cl)Cl |
| Vis mere |
Product intended for laboratory use only