Få mere at vide
Chlordiazepoxide Hydrochloride, Mikromol
Beskrivelse
Chlordiazepoxide Hydrochloride, Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chlordiazepoxide hydrochloride |
| CAS | 438-41-5 |
| Analyt- eller komponentnavne | Chlordiazepoxide hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H14 Cl N3 O . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 3H-1,4-Benzodiazepin-2-amine, 7-chloro-N-methyl-5-phenyl-, 4-oxide, hydrochloride (1:1), 3H-1,4-Benzodiazepin-2-amine, 7-chloro-N-methyl-5-phenyl-, 4-oxide, monohydrochloride (9CI), 3H-1,4-Benzodiazepine, 7-chloro-2-(methylamino)-5-phenyl-, 4-oxide, monohydrochloride (6CI,8CI), 7-Chloro-2-(methylamino)-5-phenyl-3H-1,4-benzodiazepine-4-oxide hydrochloride, A-Poxide, Ansiacal, Benzodiapin, Cebrum, Chlordiazepoxide hydrochloride, Chlordiazepoxide monohydrochloride, Chlorodiazepoxide hydrochloride, Contol, Corax, Disarim, Equibral, Labican, Lentotran, Librium, Librium hydrochloride, Methaminodiazepoxide hydrochloride, NSC 115748, OCM, Psichial, Psicoterina, Reliberan, Ro 5-0690, SK-Lygen, Seren Vita, Viansin |
| InChI formel | InChI=1S/C16H14ClN3O.ClH/c1-18-15-10-20(21)16(11-5-3-2-4-6-11)13-9-12(17)7-8-14(13)19-15;/h2-9H,10H2,1H3,(H,18,19);1H |
| Løsningstype | Neat |
| SMIL | Cl.CNC1=Nc2ccc(Cl)cc2C(=[N+]([O-])C1)c3ccccc3 |
| Vis mere |