Få mere at vide
Chlorate Standard: ClO3- @ 1000 μg/mL in H2O, VHG™
Artikelnummer. 17929064
Skift visning
Mængde:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17929064 100 ml
17928999 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17929064 Leverandør VHG Labs Leverandørnr. VHGICLO3100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Chlorate Standard: ClO3- @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chloride
CAS 16887-00-6
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Chloride
Forsendelsestilstand Room Temperature
Molekylær formel Cl-
Synonym Cl−
SMIL [Cl-]
IUPAC navn Chloride
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 35.45
Mængde 100 ml
Formel vægt 35.45 g/mol