Få mere at vide
Chlorantraniliprole-D3, TRC
Artikelnummer. 30358007
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30358007 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358007 Leverandør Toronto Research Chemicals Leverandørnr. TRCC32538425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chlorantraniliprole D3
CAS 1392493-28-5
Renhedsgrad noter HPLC
Molekylær formel C18 2H3 H11 Br Cl2 N5 O2
Mængde 25 mg
Synonym Chlorantraniliprole D3 (N-methyl D3), 1H-Pyrazole-5-carboxamide, 3-bromo-N-[4-chloro-2-methyl-6-[(methyl-d3-amino)carbonyl]phenyl]-1-(3-chloro-2-pyridinyl)-
InChI formel InChI=1S/C18H14BrCl2N5O2/c1-9-6-10(20)7-11(17(27)22-2)15(9)24-18(28)13-8-14(19)25-26(13)16-12(21)4-3-5-23-16/h3-8H,1-2H3,(H,22,27)(H,24,28)/i2D3
SMIL [2H]C([2H])([2H])NC(=O)c1cc(Cl)cc(C)c1NC(=O)c2cc(Br)nn2c3ncccc3Cl
IUPAC navn 5-bromo-N-[4-chloro-2-methyl-6-(trideuteriomethylcarbamoyl)phenyl]-2-(3-chloropyridin-2-yl)pyrazole-3-carboxamide
Molekylvægt (g/mol) 486.16
Formel vægt 483.9896
Procent renhed >95