Få mere at vide
Chloramphenicol Palmitate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chloramphenicol palmitate |
| CAS | 530-43-8 |
| Molekylær formel | C27 H42 Cl2 N2 O6 |
| Mængde | 10 g |
| Synonym | Hexadecanoic acid, (2R,3R)-2-[(dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl ester (9CI), Hexadecanoic acid, 2-[(dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl ester, [R-(R*,R*)]-, Palmitic acid, ester with chloramphenicol (6CI), Palmitic acid, α-ester with D-threo-(-)-2,2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-nitrophenethyl]acetamide (8CI), Acetamide, 2,2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-nitrophenethyl]-, α-palmitate, D-threo- (8CI), Chlorambon, Chloramphenicol monopalmitate, Chloramphenicol monopalmitate (ester), Chloramphenicol palmitate, Chloramphenicol palmitate (ester), Chloramphenicol α-palmitate, Chloromycetin palmitate, Chloropal, Clorolifarina, D-(-)-Chloramphenicol palmitate, Detreopal |
| InChI formel | InChI=1S/C27H42Cl2N2O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-24(32)37-20-23(30-27(34)26(28)29)25(33)21-16-18-22(19-17-21)31(35)36/h16-19,23,25-26,33H,2-15,20H2,1H3,(H,30,34)/t23-,25-/m1/s1 |
| SMIL | CCCCCCCCCCCCCCCC(=O)OC[C@@H](NC(=O)C(Cl)Cl)[C@H](O)c1ccc(cc1)[N+](=O)[O-] |
| IUPAC navn | [(2R,3R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl] hexadecanoate |
| Molekylvægt (g/mol) | 561.54 |
| Formel vægt | 560.242 |