missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7081875
Click to view available options
Quantity:
250 g
1000 g
5000 g
7080-50-4
C7H13ClNNaO5S
281.68
MFCD00149066
RNROOTWAJBXJBD-UHFFFAOYSA-M
benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate
122197679
N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
7080-50-4
C7H13ClNNaO5S
281.68
MFCD00149066
RNROOTWAJBXJBD-UHFFFAOYSA-M
benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate
122197679
N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

Solubility
Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.

Notes
Incompatible with strong oxidizing agents, acids and ammonia.
TRUSTED_SUSTAINABILITY
Melting Point 167°C to 170°C
Quantity 250 g
Assay Percent Range 98%
UN Number UN3263
Beilstein 3924168
Flash Point 192°C (377°F)
Merck Index 14,2075
Solubility Information Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.
Formula Weight 281.69 (227.67 Anhydrous)
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material Chloramine-T Trihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'