Learn More
Chlorambucil, LoGiCal
Product Code. 30458554
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30458554 10 mg 10mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30458554 Supplier LoGiCal Supplier No. LGCFOR0821.00
 Restricted Item
Forventet forsendelse: 30-11-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlorambucil
CAS 305-03-3
Analyte or Component Names Chlorambucil
Shipping Condition Room Temperature
Molecular Formula C14 H19 Cl2 N O2
Certifications/Compliance ISO 9001
Synonym Benzenebutanoic acid, 4-[bis(2-chloroethyl)amino]-, Butyric acid, 4-[p-[bis(2-chloroethyl)amino]phenyl]- (8CI), 4-[Bis(2-chloroethyl)amino]benzenebutanoic acid, 4-[p-[Bis(2-chloroethyl)amino]phenyl]butyric acid, Ambochlorin, Amboclorin, CB 1348, Chlorambucil, Chloraminophene, Chlorbutin, Chlorobutine, Ecloril, Leukeran, Leukeran Tablets, Linfolizin, Linfolysin, Lympholysin, NCI 3088, NSC 3088, gamma-[p-Bis(2-chloroethyl)aminophenyl]butyric acid, gamma-[p-Di(2-chloroethyl)aminophenyl]butyric acid
InChI Formula InChI=1S/C14H19Cl2NO2/c15-8-10-17(11-9-16)13-6-4-12(5-7-13)2-1-3-14(18)19/h4-7H,1-3,8-11H2,(H,18,19)
Solution Type Neat
SMILES OC(=O)CCCc1ccc(cc1)N(CCCl)CCCl
IUPAC Name 4-[4-[bis(2-chloroethyl)amino]phenyl]butanoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 304.21
Quantity 10 mg
Formula Weight 303.0793 g/mol