Få mere at vide
Chloralformamide, LoGiCal
Artikelnummer. 30462290
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30462290 5 mg
1 options
Artikelnummer. 30462290 Leverandør LoGiCal Leverandørnr. LGCFOR1862.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chloralformamide
CAS 515-82-2
Analyt- eller komponentnavne Chloralformamide
Forsendelsestilstand Room Temperature
Molekylær formel C3 H4 Cl3 N O2
Certificeringer/compliance ISO 9001
Synonym Chloralamide, Chloralformamide, Chloramide, N-(1-Hydroxy-2,2,2-trichloroethyl)formamide, N-(2,2,2-Trichloro-1-hydroxyethyl)formamide, NSC 52798, Formamide, N-(2,2,2-trichloro-1-hydroxyethyl)-
InChI formel InChI=1S/C3H4Cl3NO2/c4-3(5,6)2(9)7-1-8/h1-2,9H,(H,7,8)
Løsningstype Neat
SMIL OC(NC=O)C(Cl)(Cl)Cl
IUPAC navn N-(2,2,2-trichloro-1-hydroxyethyl)formamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 192.43
Mængde 5 mg
Formel vægt 190.9308 g/mol