missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Trachomatis Lps Antibody (1312/236), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66402H
This item is not returnable.
View return policy
Description
Chlamydia Trachomatis Lps Monoclonal antibody specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
Chlamydia Trachomatis Lps | |
Monoclonal | |
HRP | |
C. trachomatis Lipopolysaccharide;Chlamydia trachomatis Lipopolysaccharide | |
Native | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA | |
1312/236 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chlamydia Trachomatis Lps Antibody (1312/236), HRP, Novus Biologicals™ > 0.1 mL; HRP
Spot an opportunity for improvement?Share a Content Correction